4-Methylpiperidine 99%

Danger

AC12751-1000 AC12751-5000 AC12751-0025
103525-314EA 509 USD
103525-314 200010-562 200010-560
4-Methylpiperidine 99%
4-Methylpiperidine
Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 122 °C (760 mmHg)
Melting Pt: 4…5 °C
Density: 0.84 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00006005
CAS Number: 626-58-4
EINECS: 210-954-1
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4480 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR