2-Methylpiperidine 99%

Danger

AC12749-1000 AC12749-5000
200022-838EA 32 USD
200022-838 200047-034
2-Methylpiperidine 99%
2-Methylpiperidine
Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 5…4 °C
Density: 0.844 g/cm³
Flash Pt: 16 °C
MDL Number: MFCD00005982
CAS Number: 109-05-7
EINECS: 203-642-1
UN: 2733

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4459 to 1.4479 (20 °C, 589 nm)
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR