2-Methylnaphthalene 96%

Warning

Synonyms: β-Methylnaphthalene

AC12717-5000 AC12717-0025 AC12717-0050
103525-304EA 206 USD
103525-304 200016-698 200009-522
2-Methylnaphthalene 96%
2-Methylnaphthalene
Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 241…242 °C (760 mmHg)
Melting Pt: 32…36 °C
Density: 1.01 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00004118
CAS Number: 91-57-6
EINECS: 202-078-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Melting point 32 to 36 °C
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR