1-Methylnaphthalene 96%

Warning

Synonyms: α-Methylnaphthalene

AC12716-1000 AC12716-5000 AC12716-0025
103525-302EA 396 USD
103525-302 200000-684 200008-726
1-Methylnaphthalene 96%
1-Methylnaphthalene
Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 240…243 °C (760 mmHg)
Melting Pt: –22 °C
Density: 1.02 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00004034
CAS Number: 90-12-0
EINECS: 201-966-8
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.6100 to 1.6180 (20 °C, 589 nm)
GC ≥95 %
GC ≥99.4 %
Color scale ≤10 APHA
Aldehyde (C3H6O) ≤0.01

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR