2-Methylimidazole 99%

Danger

Synonyms: 2-Methyl-1H-imidazole

AC12700-1000 AC12700-5000 AC12700-0010
200022-832EA 73.2 USD
200022-832 200028-876 200005-118
2-Methylimidazole 99%
2-Methylimidazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 142…145 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00005190
CAS Number: 693-98-1
EINECS: 211-765-7
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 142 to 145 °C
Water ≤0.5 % (K.F.)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR