Methyl formate 97%, pure

Danger

Synonyms: Formic acid methyl ester

AC12682-0025
200000-664EA 125 USD
200000-664
Methyl formate 97%, pure
Methyl formate
Formula: C₂H₄O₂
MW: 60.05 g/mol
Boiling Pt: 31…33 °C (760 mmHg)
Melting Pt: –100 °C
Density: 0.968 g/cm³
Flash Pt: –32 °C
Storage Temperature: Ambient
MDL Number: MFCD00003291
CAS Number: 107-31-3
EINECS: 203-481-7
UN: 1243
ADR: 3,I
Merck Index: 12,06153

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3420 to 1.3440 (20 °C, 589 nm)
Water ≤0.05 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR