2,2'-Methyliminodiethanol 99+%
|
Warning
|
Synonyms:
AMIETOL M 12
, MDEA
, N-Methyldiethanolamine
, N,N-Diethanolmethylamine
, N-Methyl-2,2'-iminodiethanol
AC12672-0010
200020-108EA
59.6
USD
200020-108
2,2'-Methyliminodiethanol 99+%
2,2'-Methyliminodiethanol
Formula:
H₃CN(CH₂CH₂OH)₂ MW: 119.16 g/mol Boiling Pt: 247 °C (1013 hPa) Melting Pt: –21 °C Density: 1.038 g/cm³ Flash Pt: 138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002848 CAS Number: 105-59-9 EINECS: 203-312-7 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4675 to 1.4695 (20 °C, 589 nm) |
Water | ≤0.5 % |
GC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...