2-Methyl-1,3-cyclohexanedione 98+%

Warning

Synonyms: 2-Methylcyclohexane-1,3-dione

AC12659-2000 AC12659-0250 AC12659-1000
200041-146EA 292 USD
200041-146 200022-782 103464-752
2-Methyl-1,3-cyclohexanedione 98+%
2-Methyl-1,3-cyclohexanedione
Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Melting Pt: >204 °C
Storage Temperature: Ambient
MDL Number: MFCD00001587
CAS Number: 1193-55-1
EINECS: 212-153-2

Order Now

Specification Test Results

Appearance Off-white to pink-orange to light brown Powder or crystals
Infrared spectrum Conforms
Melting point ≥204 °C
Water ≤1 % (K.F.)
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR