Isoamyl alcohol 98%, pure

Warning

Synonyms: 3-Methyl-1-butanolIsopentyl alcohol3-Methylbutanoliso-Pentanol

AC12648-0010 AC12648-0025 AC12648-5000 AC12648-0250
200000-660EA 84 USD
200000-660 200000-658 103525-280 200017-140
Isoamyl alcohol 98%, pure
Isoamyl alcohol
Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 130 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.8 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Merck Index: 13,05214
https://us.vwr.com/stibo/web/std.lang.all/15/69/2301569.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20 °C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR