Methyl-5-bromovalerate 97%

Synonyms: 5-Bromovaleric acid methyl esterMethyl 5-Bromovalerate

AC12646-0100 AC12646-0250
200037-928EA 140 USD
200037-928 200019-708
Methyl-5-bromovalerate 97%
Methyl 5-bromovalerate
Formula: BrH₂C(CH₂)₃COOCH₃
MW: 195.06 g/mol
Boiling Pt: 79…80 °C (5.33 hPa)
Density: 1.36 g/cm³
Flash Pt: 99 °C
MDL Number: MFCD00000265
CAS Number: 5454-83-1
EINECS: 226-709-7

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4640 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR