9-Methylanthracene 99%

Danger

AC12630-0500 AC12630-0100 AC12630-0050
200036-430EA 61.7 USD
200036-430 103525-276 200017-948
9-Methylanthracene 99%
9-Methylanthracene
Formula: C₁₅H₁₂
MW: 192.26 g/mol
Boiling Pt: 196…197 °C (12 mmHg)
Melting Pt: 76…79 °C
Density: 1.066 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00001261
CAS Number: 779-02-2
EINECS: 212-299-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 77 to 80 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR