2-Methylanisole 99%

Warning

Synonyms: 2-Methoxytolueneo-Cresol methyll ether2-Methylanisole1-Methoxy-2-methylbenzene

AC12628-5000 AC12628-0050 AC12628-1000
200034-910EA 89.2 USD
200034-910 200047-000 200028-868
2-Methylanisole 99%
2-Methylanisole
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 172 °C (1013 hPa)
Density: 0.98 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00008373
CAS Number: 578-58-5
EINECS: 209-426-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5180 (20 °C, 589 nm)
GC ≥98.5 %
Residue after evaporation ≤0.0005 %
Acidity ≤0.0005 meq/g
Free chlorine ≤0.00003 % (as Cl)
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Aldehyde (HCHO) ≤0.0005 %
Ethanol ≤0.3 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR