Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methylaniline ≥99%

Supplier: ACROS ORGANICS MS

Danger

AC12627-0010 AC12627-5000 AC12627-0025
200013-690EA 73.86 USD InStock
200013-690 200007-186 103525-274
N-Methylaniline ≥99%
N-Methylaniline


Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 195 °C (760 mmHg)
Melting Pt: –57 °C
Density: 0.989 g/cm³ (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00008283
CAS Number: 100-61-8
EINECS: 202-870-9
UN: 2294
ADR: 6.1,III
Merck Index: 12,06098
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5694 to 1.5714 (20°C, 589 nm)
GC ≥98.5 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty