2'-Methylacetophenone 98%

Synonyms: Methyl o-tolyl ketone1-(o-Tolyl)ethanoneo-Methylacetophenone

AC12616-1000 AC12616-0050 AC12616-0250
103464-816EA 202 USD
103464-816 103464-814 200016-676
2'-Methylacetophenone 98%
2'-Methylacetophenone
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 214 °C (760 mmHg)
Density: 1.026 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00008734
CAS Number: 577-16-2
EINECS: 209-408-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5330 (20 °C, 589 nm)
GC ≥97.5 %
UV transmittance ≥99 % (c=0.01M water, 254 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR