1-(2-Methoxyphenyl)piperazine 98+%

Danger

AC12604-1000 AC12604-0250 AC12604-0050
200008-320EA 30 USD
200008-320 200027-472 103525-270
1-(2-Methoxyphenyl)piperazine 98+%
1-(2-Methoxyphenyl)piperazine
Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
Boiling Pt: 130…133 °C (.1 mmHg)
Melting Pt: 35…38 °C
Density: 1.09 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005958
CAS Number: 35386-24-4
EINECS: 252-537-7
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5745 to 1.5765 (20 °C, 589 nm)
Titration Tertiary base 98.0 to 102.0 %
GC ≥98.0 %
GC ≤1.0 % (o-anisidine)
Titration with HClO4 98.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR