Bis(2-methoxyethyl)ether 99%, Extra Pure

Danger

Synonyms: 2-Methoxyethyl etherDiethylene glycol dimethyl ether1-Methoxy-2-(2-methoxyethoxy)ethaneDiglymeDimethyldigol

AC12585-2500 AC12585-0010 AC12585-0100 AC12585-0025
103525-268EA 545.77 USD
103525-268 200000-636 200000-634 200009-464
Bis(2-methoxyethyl)ether 99%, Extra Pure
Bis(2-methoxyethyl) ether
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 162 °C (760 mmHg)
Melting Pt: –64 °C
Density: 0.937 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00008503
CAS Number: 111-96-6
EINECS: 203-924-4
UN: 3271
ADR: 3,III
Merck Index: 13,03187

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4090 (20 °C, 589 nm)
Water ≤0.025 %
GC ≥98.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Acidity (CH3COOH) ≤0.015 %
Peroxides ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR