3-Methoxy-1-butanol 99%

Warning

Synonyms: 3-Methoxybutan-1-ol

AC12579-1000 AC12579-0010
200041-614EA 20.2 USD
200041-614 200035-446
3-Methoxy-1-butanol 99%
3-Methoxy-1-butanol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 161 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.92 g/cm³
Flash Pt: 46 °C
MDL Number: MFCD00002931
CAS Number: 2517-43-3
EINECS: 219-741-8
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4165 (20 °C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR