2,4,6-Trimethylbenzaldehyde 97%

Warning

Synonyms: Mesitaldehyde

AC12557-0100 AC12557-0500
200019-622EA 82.3 USD
200019-622 200025-768
2,4,6-Trimethylbenzaldehyde 97%
2,4,6-Trimethylbenzaldehyde
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 14 °C
Density: 1 g/cm³
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00003341
CAS Number: 487-68-3
EINECS: 207-662-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5520 to 1.5540 (20 °C, 589 nm)
GC ≥96.0 %
Loss on ignition 24.5 % to 26.5 % (800 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR