(R)-(-)-Mandelic acid 99%

Danger

Synonyms: (R)-(-)-2-Hydroxy-2-phenylacetic acid(R)-2-Hydroxy-2-phenylacetic acidD(-)-2-Hydroxy-2-phenylacetic acidD(-)-Mandelic acid

AC12532-1000 AC12532-0050 AC12532-0250
200014-326EA 52 USD
200014-326 200032-744 103508-686
(R)-(-)-Mandelic acid 99%
(R)-(-)-Mandelic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 130…135 °C
Flash Pt: >190 °C
Storage Temperature: Ambient
MDL Number: MFCD00064251
CAS Number: 611-71-2
EINECS: 210-276-6

Order Now

Specification Test Results

Appearance White to light yellow to light beige Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 130 to 135 °C
Water ≤1 % (K.F.)
Sulfated ash ≤0.2 %
Specific optical rotation -152° to -158° (20 °C, 589 nm) (c=2, H2O)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR