2,6-Lutidine 99%

Warning

Synonyms: 2,6-Dimethylpyridine

AC12519-0025 AC12519-2500
103525-232EA 265 USD
103525-232 200000-602
2,6-Lutidine 99%
2,6-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 143…145 °C (760 mmHg)
Melting Pt: –6 °C
Density: 0.92 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00006345
CAS Number: 108-48-5
EINECS: 203-587-3
UN: 1993
ADR: 3,III
Merck Index: 13,05636

Order Now

Specification Test Results

Appearance Clear liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4966 to 1.4986 (20 °C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR