N-(2-Hydroxyethyl)phthalimide 99%

Synonyms: 2-(2-Hydroxyethyl)isoindoline-1,3-dione

AC12492-1000 AC12492-0250 AC12492-5000
200025-986EA 24 USD
200025-986 103508-604 200038-008
N-(2-Hydroxyethyl)phthalimide 99%
N-(2-Hydroxyethyl)phthalimide
Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00005903
CAS Number: 3891-07-4
EINECS: 223-434-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 129 °C
HPLC ≥98.5 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR