Octadecylbenzene 98%

Synonyms: 1-Phenyloctadecane

AC12470-0010
200032-372EA 102 USD
200032-372
Octadecylbenzene 98%
Octadecylbenzene
Formula: C₂₄H₄₂
MW: 330.6 g/mol
Boiling Pt: 249 °C (15 mmHg)
Melting Pt: 32…36 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00048500
CAS Number: 4445-07-2
EINECS: 224-684-7

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 32 to 36 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR