Aluminium sulfate octadecahydrate, Extra Pure

Danger

Synonyms: Aluminum(III) sulfate octadecahydrateAluminum(III) sulphate octadecahydrate

AC12368-0025
200048-532EA 180 USD
200048-532
Aluminium sulfate octadecahydrate, Extra Pure
Aluminium sulphate octadecahydrate
Formula: Al₂(SO₄)₃·18H₂O
MW: 666.43 g/mol
Melting Pt: 86 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149136
CAS Number: 7784-31-8
EINECS: 233-135-0
https://us.vwr.com/stibo/web/std.lang.all/09/26/18570926.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric 98.0 to 110.0 %
Chloride (Cl) ≤50 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR