Isobutyraldehyde 99+%

Danger

Synonyms: 2-Methylpropionaldehyde2-Methylpropanal

AC12251-0025 AC12251-0100 AC12251-0010
200000-572EA 47 USD
200000-572 200019-388 103525-164
Isobutyraldehyde 99+%
Isobutyraldehyde
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 63 °C (760 mmHg)
Melting Pt: –65 °C
Density: 0.79 g/cm³
Flash Pt: –24 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006980
CAS Number: 78-84-2
EINECS: 201-149-6
UN: 2045
ADR: 3,II
Merck Index: 13,05171

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3710 to 1.3745 (20 °C, 589 nm)
GC ≥99.0 %
Color scale ≤10 APHA
Free acid ≤1.0 % (Isobutyric acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR