4-Iodophenol 99%

Danger

AC12239-0100 AC12239-0250 AC12239-1000 AC12239-5000
200010-496EA 79.9 USD
200010-496 103525-162 200005-470 103525-160
4-Iodophenol 99%
4-Iodophenol
Formula: C₆H₅IO
MW: 220.01 g/mol
Boiling Pt: 287 °C (760 mmHg)
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00002327
CAS Number: 540-38-5
EINECS: 208-745-5
UN: 3261
ADR: 8,III
Merck Index: 13,05058

Order Now

Specification Test Results

Appearance Beige to pink to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 89 to 94 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR