2-Iodobenzoic acid 98%

Danger

Synonyms: o-Iodobenzoic acid

AC12234-5000 AC12234-0250 AC12234-1000
200000-568EA 64.8 USD
200000-568 200009-440 103463-540
2-Iodobenzoic acid 98%
2-Iodobenzoic acid
Formula: C₇H₅IO₂
MW: 248.02 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00002419
CAS Number: 88-67-5
EINECS: 201-850-7
Merck Index: 13,05052

Order Now

Specification Test Results

Appearance White to off-white to beige Powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure
Calcium (Ca) ≤0.003 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.01 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR