Iodobenzene 98%

Warning

Synonyms: Phenyliodide

AC12233-1000 AC12233-0025 AC12233-5000
200019-372EA 316.62 USD
200019-372 200000-566 103510-048
Iodobenzene 98%
Iodobenzene
Formula: C₆H₅I
MW: 204.01 g/mol
Boiling Pt: 188 °C (760 mmHg)
Melting Pt: –29 °C
Density: 1.82 g/cm³
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00001029
CAS Number: 591-50-4
EINECS: 209-719-6
Merck Index: 13,05051

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6180 to 1.6200 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR