4-Iodoanisole 98%

AC12232-5000 AC12232-1000 AC12232-0250
200005-466EA 35.7 USD
200005-466 103510-046 200017-592
4-Iodoanisole 98%
4-Iodoanisole
Formula: C₇H₇IO
MW: 234.04 g/mol
Boiling Pt: 237 °C (726 mmHg)
Melting Pt: 46…51 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001056
CAS Number: 696-62-8
EINECS: 211-798-7

Order Now

Specification Test Results

Appearance Off-white to brown Crystalline powder and/or chunks (May darken on storage)
Infrared spectrum Conforms
Melting point 46 to 51 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR