Iodoacetonitrile 95%

Danger

Synonyms: 2-Iodoacetonitrile

AC12229-0050 AC12229-0250
103530-750EA 515 USD
103530-750 200012-776
Iodoacetonitrile 95%
Iodoacetonitrile
Formula: C₂H₂IN
MW: 166.95 g/mol
Boiling Pt: 182…184 °C (720 mmHg)
Density: 2.3 g/cm³
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001886
CAS Number: 624-75-9
EINECS: 210-860-0
UN: 3276
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5740 to 1.5780 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR