Iodoacetamide 98%

Danger

Synonyms: 2-Iodoacetamide

AC12227-5000 AC12227-0050 AC12227-1000 AC12227-0250
200000-562EA 49.2 USD
200000-562 200000-564 200012-766 103525-150
Iodoacetamide 98%
Iodoacetamide
Formula: ICH₂CONH₂
MW: 184.96 g/mol
Melting Pt: >90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point ≥90 °C
Titration Argentometric ≥97.5 % (ex I)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR