Indole 99+%

Danger

Synonyms: 1H-Indole

AC12215-0025 AC12215-0250 AC12215-0100 AC12215-1000
103525-148EA 795 USD
103525-148 200006-692 200012-734 200012-736
Indole 99+%
Indole
Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 253…254 °C (760 mmHg)
Melting Pt: 51…53 °C
Density: 1.22 g/cm³
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,04985

Order Now

Specification Test Results

Appearance White to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 51 to 53 °C
GC ≥99.0 %
UV at 400 nm A: ≤0.01
UV at 240 nm A: ≤0.01
UV at 220 nm A: ≤0.07
UV at 210 nm A: ≤0.30
UV at 195 nm A: ≤1.0
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR