1,3-Indandione 97%

Warning

Synonyms: 1H-Indene-1,3(2H)-dioneIndan-1,3-dione1,3-Indanedione

AC12209-0250 AC12209-1000 AC12209-0050
103510-022EA 258 USD
103510-022 200027-674 200045-762
1,3-Indandione 97%
1,3-Indandione
Formula: C₉H₆O₂
MW: 146.15 g/mol
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00003779
CAS Number: 606-23-5
EINECS: 210-109-7

Order Now

Specification Test Results

Appearance Light yellow to beige to golden Powder
Infrared spectrum Conforms
Melting point 124 to 131 °C
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR