Hypoxanthine 99.5%

Warning

Synonyms: 1,9-Dihydro-6H-purin-6-one1H-Purin-6-onePurin-6(1H)-one6-Purinone

AC12201-0050 AC12201-0250 AC12201-1000
200000-552EA 91.71 USD
200000-552 103510-020 103510-018
Hypoxanthine 99.5%
Hypoxanthine
Formula: C₅H₄N₄O
MW: 136.11 g/mol
Melting Pt: >250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005725
CAS Number: 68-94-0
EINECS: 200-697-3
Merck Index: 13,04895

Order Now

Specification Test Results

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Sulfated ash ≤0.2 %
HPLC ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR