4-Quinolinol 98%

Warning

Synonyms: 4-HydroxyquinolineQuinolin-4-ol

AC12189-0500 AC12189-0100
103530-748EA 757 USD
103530-748 200010-480
4-Quinolinol 98%
4-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 202…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00006777
CAS Number: 611-36-9
EINECS: 210-268-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to light beige Crystalline powder or needles
Infrared spectrum Conforms
Melting point 202 to 206 °C
Titration with HClO4 ≥97.5 %
Specific optical rotation +9.5° to +10.5° (20 °C, 589 nm) (c=4 in H2O) reading after >1 hour
Iron (Fe) ≤0.2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR