2-Methyl-8-quinolinol ≥98%
|
Warning
|
Synonyms:
8-Hydroxyquinaldine
, 8-Hydroxy-2-methylquinoline
, 2-Methyloxine
AC12187-5000
AC12187-0250
200030-218EA
148.5
USD
200030-218
200024-236
2-Methyl-8-quinolinol ≥98%
2-Methyl-8-quinolinol
Formula:
C₁₀H₉NO MW: 159.19 g/mol Boiling Pt: 267 °C (1013 hPa) Melting Pt: 70…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006765 CAS Number: 826-81-3 EINECS: 212-562-6 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | Beige to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 68 to 73 °C |
Titration with HClO4 | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...