4-Hydroxy-1-methylpiperidine 98%

Warning

Synonyms: 1-Methylpiperidin-4-ol1-Methyl-4-piperidinolN-Methyl-4-piperidinol

AC12151-5000 AC12151-1000 AC12151-0050
200029-854EA 56.32 USD
200029-854 200010-450 200042-052
4-Hydroxy-1-methylpiperidine 98%
4-Hydroxy-1-methylpiperidine
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 29…31 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00006500
CAS Number: 106-52-5
EINECS: 203-406-8

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4790 (20 °C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.1 ppm
Potassium (K) ≤0.2 ppm
Sodium (Na) ≤1 ppm
Zinc (Zn) ≤0.1 ppm
Total phosphorus (P) ≤0.5 ppm
Total silicon (Si) ≤0.02 ppm
Total sulfur (S) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR