3-Hydroxy-3-methylbutanone 92%

Warning

Synonyms: 3-Hydroxy-3-methyl-2-butanone3-Hydroxy-3-methylbutan-2-one

AC12144-0250 AC12144-0050
103530-738EA 59.88 USD
103530-738 200035-418
3-Hydroxy-3-methylbutanone 92%
3-Hydroxy-3-methylbutanone
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 140…141 °C (760 mmHg)
Density: 0.97 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00004460
CAS Number: 115-22-0
EINECS: 204-073-1
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light red Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4170 (20 °C, 589 nm)
GC ≥90 %
UV 1 cm cell vs HPLC-grade water
UV at 200 nm A: ≤0.3
UV at 210 nm A: ≤0.15
UV at 220 nm A: ≤0.07
UV at 230 nm A: ≤0.03
UV at 240 nm A: ≤0.02
Residue after evaporation ≤2 ppm
Acidity ≤0.00015 meq/g
Filtered to 0.2 micrometer Filtered

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR