Diethylene glycol 99%, Extra Pure

Warning

Synonyms: Digol2,2'-Oxybisethanol2,2'-Oxybis(ethan-1-ol)

AC12116-0025 AC12116-0010 AC12116-0100
200000-544EA 44 USD
200000-544 200000-546 103509-598
Diethylene glycol 99%, Extra Pure
Diethylene glycol
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 245 °C (760 mmHg)
Melting Pt: –10 °C
Density: 1.11 g/cm³
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00002882
CAS Number: 111-46-6
EINECS: 203-872-2
Merck Index: 13,03146
https://us.vwr.com/stibo/web/std.lang.all/77/97/2297797.jpg

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4480 (20 °C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Color scale ≤20 APHA
Ash ≤0.01 %
Acidity (CH3COOH) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR