(±)-3-Hydroxybutyric acid 95%, Technical Grade

Warning

Synonyms: (±)-3-Hydroxybutanoic acid3-Hydroxybutyric acidDL-3-Hydroxybutyric Acid

AC12105-0500 AC12105-0050
103509-596EA 903 USD
103509-596 200017-098
(±)-3-Hydroxybutyric acid 95%, Technical Grade
(±)-3-Hydroxybutyric acid
Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 118…120 °C (2 mmHg)
Flash Pt: 112 °C
Storage Temperature: Freezer
MDL Number: MFCD00004546
CAS Number: 300-85-6
EINECS: 206-099-9
Merck Index: 12,04861

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Solid or liquid
Infrared spectrum Conforms
Titration with NaOH 94 to 106 %
HPLC ≥98.5 %
Specific optical rotation +80° to +86° (20 °C, 589 nm) (c=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR