Hexamethylbenzene 98+%

AC12057-0250 AC12057-0050
200012-656EA 190 USD
200012-656 103509-568
Hexamethylbenzene 98+%
Hexamethylbenzene
Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 264 °C (760 mmHg)
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00008523
CAS Number: 87-85-4
EINECS: 201-777-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 164 to 168 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR