Sorbic acid 99%

Warning

Synonyms: (E,E)-2,4-Hexadienoic acid2,4-Hexadienoic acid(2E,4E)-2,4-Hexadienoic acidtrans,trans-2,4-Hexadienoic acid(2E,4E)-Hexa-2,4-dienoic acid

AC12053-0010 AC12053-2500 AC12053-1000 AC12053-5000
200009-006EA 26.3 USD
200009-006 200009-008 200009-010 200028-292
Sorbic acid 99%
Sorbic acid
Formula: CH₃CH=CHCH=CHCOOH
MW: 112.13 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 132…135 °C
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002703
CAS Number: 110-44-1
EINECS: 203-768-7
Merck Index: 12,08869
https://us.vwr.com/stibo/web/std.lang.all/29/27/2302927.jpg

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 132 to 135 °C
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.2 %
Titration with NaOH 98.5 to 101.5 % (On anhydrous substance)
Arsenic (As) ≤3 ppm
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR