1-Hexadecene 92% tech.

Danger

AC12050-0010
200004-744EA 55.1 USD
200004-744
1-Hexadecene 92% tech.
1-Hexadecene
Formula: C₁₆H₃₂
MW: 224.43 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 3…5 °C
Density: 0.783 g/cm³
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00008991
CAS Number: 629-73-2
EINECS: 211-105-8

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4430 (20 °C, 589 nm)
GC ≥90 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR