1-Heptanol 98%

Warning

Synonyms: Heptyl alcohol

AC12036-0025 AC12036-0010 AC12036-2500
200004-740EA 32.2 USD
200004-740 103509-504 200008-676
1-Heptanol 98%
1-Heptanol
Formula: H₃C(CH₂)₅CH₂OH
MW: 116.2 g/mol
Boiling Pt: 176 °C (760 mmHg)
Melting Pt: –35 °C
Density: 0.8219 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002986
CAS Number: 111-70-6
EINECS: 203-897-9
Merck Index: 13,04681

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4250 (20 °C, 589 nm)
Water ≤0.5 %
GC ≥97.5 %
Color scale ≤30 APHA
Residue after evaporation ≤0.01 %
Bromine number ≤0.1 g Br/100 g
Acid number ≤0.5 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR