Guanidinium carbonate 99+%

Warning

Synonyms: Guanidine carbonateGuanidine carbonate saltDiguanidinium carbonate

AC12022-2500 AC12022-0010 AC12022-0051
200031-492EA 77 USD
200031-492 103516-426 200006-600
Guanidinium carbonate 99+%
Guanidinium carbonate
Formula: C₃H₁₂N₆O₃
MW: 180.17 g/mol
Melting Pt: 267…270 °C
Storage Temperature: Ambient
MDL Number: MFCD00013029
CAS Number: 593-85-1
EINECS: 209-813-7

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %
HPLC ≥97.5 %
pH 10.5 to 11.5 (1 % in water)
Specific optical rotation -9° to -14.5° (20 °C, 589 nm) (c=4 in H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR