Guaiacol 99+%

Warning

Synonyms: 2-Methoxyphenol2-Hydroxyanisole1-Hydroxy-2-methoxybenzene

AC12019-0050 AC12019-0010 AC12019-2500
200000-500EA 48.29 USD
200000-500 103509-494 200019-276
Guaiacol 99+%
Guaiacol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 205 °C (760 mmHg)
Melting Pt: 27…29 °C
Density: 1.12 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00002185
CAS Number: 90-05-1
EINECS: 201-964-7
ADR: 6.1,III
Merck Index: 13,04568

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or crystalline mass
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5450 (20 °C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR