Glutaric anhydride 95%

Danger

Synonyms: Dihydro-2H-pyran-2,6(3H)-dione

AC11997-1000 AC11997-5000 AC11997-0050
200012-588EA 34.9 USD
200012-588 200006-578 200012-590
Glutaric anhydride 95%
Glutaric anhydride
Formula: C₅H₆O₃
MW: 114.1 g/mol
Boiling Pt: 290 °C (760 mmHg)
Melting Pt: 46…57 °C
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00006679
CAS Number: 108-55-4
EINECS: 203-593-6

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder or crystals and/or chunks
Infrared spectrum Conforms
GC ≥94 %
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR