Glutaric acid 99%

Warning

Synonyms: 1,5-Pentanedioic acid

AC11996-5000 AC11996-0250 AC11996-1000
200012-584EA 61.3 USD
200012-584 200012-586 200031-468
Glutaric acid 99%
Glutaric acid
Formula: C₅H₈O₄
MW: 132.12 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 92…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00004410
CAS Number: 110-94-1
EINECS: 203-817-2
Merck Index: 13,04486

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 95 to 99 °C
Titration with NaOH 98.5 to 101.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR