Fumaric acid 99+%

Warning

AC11975-0100 AC11975-0010 AC11975-1000 AC11975-0025
200012-544EA 84 USD
200012-544 103508-822 103508-824 200006-562
Fumaric acid 99+%
Fumaric acid
Formula: C₄H₄O₄
MW: 116.07 g/mol
Melting Pt: 295…300 °C (subl.)
Density: 1.62 g/cm³
Flash Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00002700
CAS Number: 110-17-8
EINECS: 203-743-0
Merck Index: 13,04308

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 295 to 300 °C
Water ≤0.25 % (K.F.)
HPLC ≥99.0 %
Titration with NaOH 99.0 to 101.0 % (On anhydrous substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR