4-Fluorotoluene 97%

Danger

Synonyms: 1-Fluoro-4-methylbenzene

AC11967-0100 AC11967-1000 AC11967-2500
200044-408EA 23.9 USD
200044-408 200032-496 200038-468
4-Fluorotoluene 97%
4-Fluorotoluene
Formula: C₇H₇F
MW: 110.13 g/mol
Boiling Pt: 116 °C (1013 hPa)
Melting Pt: –56 °C
Density: 1 g/cm³
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000358
CAS Number: 352-32-9
EINECS: 206-520-6
UN: 2388
ADR: 3,II
Merck Index: 13,04207

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4674 to 1.4694 (20 °C, 589 nm)
Water ≤0.05 % (K.F.)
GC ≥96.0 %
Specific optical rotation +9.5° to +11.5° (20 °C, 589 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR