4-Fluorophenol 99%

Warning

Synonyms: p-Fluorophenol1-Fluoro-4-hydroxybenzene

AC11956-1000 AC11956-0250
200010-432EA 40 USD
200010-432 200017-544
4-Fluorophenol 99%
4-Fluorophenol
Formula: C₆H₅FO
MW: 112.1 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 44…49 °C
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002316
CAS Number: 371-41-5
EINECS: 206-736-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to beige to pink Crystalline solid
Infrared spectrum Conforms
Melting point 44 to 49 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR